Learn More
2',5'-Dimethoxyacetophenone, TRC
Product Code. 30382851
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382851 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30382851 Supplier Toronto Research Chemicals Supplier No. TRCD5621005G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2',5'-Dimethoxyacetophenone
CAS 1201-38-3
Molecular Formula C10H12O3
Quantity 5 g
Synonym 1-(2,5-dimethoxyphenyl)-Ethanone, Acetophenone, 2',5'-dimethoxy- (6CI,7CI,8CI), 1-(2,5-Dimethoxyphenyl)ethanone, Methyl 2,5-dimethoxyphenyl ketone, NSC 62094
InChI Formula InChI=1S/C10H12O3/c1-7(11)9-6-8(12-2)4-5-10(9)13-3/h4-6H,1-3H3
SMILES COc1ccc(OC)c(c1)C(=O)C
Molecular Weight (g/mol) 180.2
Formula Weight 180.0786