Learn More
2,5-Diformylfuran, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Diformylfuran |
| CAS | 823-82-5 |
| Molecular Formula | C6 H4 O3 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C6H4O3/c7-3-5-1-2-6(4-8)9-5/h1-4H |
| SMILES | O=Cc1oc(C=O)cc1 |
| IUPAC Name | furan-2,5-dicarbaldehyde |
| Molecular Weight (g/mol) | 124.09 |
| Formula Weight | 124.016 |