missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroanisole, 98%

Product Code. p-7046221
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353107 5 g 5g
11363107 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353107 Supplier Thermo Scientific Alfa Aesar Supplier No. L13497.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1984-58-3 | C7H6Cl2O | 177.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-58-3
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00061123
InChI Key QKMNFFSBZRGHDJ-UHFFFAOYSA-N
Synonym 2,5-dichloroanisole, benzene, 1,4-dichloro-2-methoxy, 2,5-dichloromethoxybenzene, banair, anisole, 2,5-dichloro, unii-8b3j0y3cj1, 2,5-dichloroanizole, pubchem3681, dsstox_cid_7452
PubChem CID 16125
IUPAC Name 1,4-dichloro-2-methoxybenzene
SMILES COC1=CC(Cl)=CC=C1Cl

Specifications

Density 1.33
Boiling Point 128°C to 132°C (3.5 mmHg)
Refractive Index 1.563
Quantity 5 g
Beilstein 2327401
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichloroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.