Learn More
2,5-Diaminotoluene (>80%), TRC
Product Code. 30135523
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135523 10 g 10g
30132418 5 g 5g
18080730 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135523 Supplier Toronto Research Chemicals Supplier No. TRCT53587710G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-70-5
Molecular Formula C7 H10 N2
Molecular Weight (g/mol) 122.17
Synonym Toluene-2,5-diamine (7CI,8CI), 2-Methyl-1,4-benzenediamine, 1,4-Diamino-2-methylbenzene, 1-Methyl-2,5-diaminobenzene, 2,5-Diaminotoluene, 2,5-Diaminotoluol, 2-Methyl-1,4-phenylenediamine, 2-Methyl-p-phenylenediamine, 4-Amino-2-methylaniline, 4-Amino-3-methylaniline, C.I. 76042, Toluylene-2,5-diamine, p-Toluenediamine
IUPAC Name 2-methylbenzene-1,4-diamine
SMILES Cc1cc(N)ccc1N
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H10N2/c1-5-4-6(8)2-3-7(5)9/h2-4H,8-9H2,1H3
Formula Weight 122.0844
Chemical Name or Material 2,5-Diaminotoluene