Learn More
2-(4-Pyridyl)ethanesulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4-Pyridyl)ethanesulfonic Acid |
| CAS | 53054-76-5 |
| Molecular Formula | C7H9NO3S |
| Quantity | 1 g |
| Synonym | 4-(2-Sulfoethyl)pyridine, 4-Pyridylethylsulfonic Acid, NSC 87853, gamma-(2-Sulfoethyl)pyridine |
| InChI Formula | InChI=1S/C7H9NO3S/c9-12(10,11)6-3-7-1-4-8-5-2-7/h1-2,4-5H,3,6H2,(H,9,10,11) |
| SMILES | OS(=O)(=O)CCc1ccncc1 |
| IUPAC Name | 2-pyridin-4-ylethanesulfonic acid |
| Molecular Weight (g/mol) | 187.22 |
| Formula Weight | 187.0303 |