missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)thiazole-4-acetic acid, 97%, Thermo Scientific™

Product Code. 15490918
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490918 5 g 5g
15481516 250 mg 250mg
15480918 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490918 Supplier Thermo Scientific Alfa Aesar Supplier No. H54713.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23353-14-2
Molecular Formula C12H11NO3S
Molecular Weight (g/mol) 249.284
MDL Number MFCD01316979
InChI Key HWRQFRXTBPYUEK-UHFFFAOYSA-N
PubChem CID 738083
IUPAC Name 2-[2-(4-methoxyphenyl)-1,3-thiazol-4-yl]acetic acid
SMILES COC1=CC=C(C=C1)C2=NC(=CS2)CC(=O)O

Specifications

Quantity 5 g
Beilstein 960002
Synonym 2-2-4-methoxyphenyl-1,3-thiazol-4-yl acetic acid, 2-4-methoxyphenyl-1,3-thiazol-4-yl acetic acid, 2-2-4-methoxyphenyl thiazol-4-yl acetic acid, 4-thiazoleacetic acid, 2-4-methoxyphenyl, 2-2-4-methoxyphenyl-1,3-thiazol-4-yl-acetic acid, 2-4-methoxyphenyl-4-thiazoleacetic acid, methoxyphenylthiazolylaceticacid, 4-thiazoleacetic acid,2-4-methoxyphenyl
Formula Weight 249.28
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxyphenyl)thiazole-4-acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.