Learn More
2,4-Dimethylbenzenethiol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 13616-82-5 |
| Molecular Formula | C8H10S |
| MDL Number | MFCD00010019 |
| Quantity | 5 g |
| SMILES | SC1=CC=C(C)C=C1C |
| Molecular Weight (g/mol) | 138.23 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4-Dimethylbenzenethiol |