Learn More
2,4'-Dihydroxydiphenylmethane, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4'-Dihydroxydiphenylmethane |
| CAS | 2467-03-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13H12O2 |
| Quantity | 250 mg |
| Synonym | 2-[(4-Hydroxyphenyl)methyl]phenol, (2-Hydroxyphenyl)(4-hydroxyphenyl)methane, 2,4'-Bis(hydroxyphenyl)methane, 2,4'-Methylenebisphenol, 2,4'-Methylenediphenol, NSC 36395, o,p'-Bis(hydroxyphenyl)methane, o-(p-Hydroxybenzyl)phenol |
| InChI Formula | InChI=1S/C13H12O2/c14-12-7-5-10(6-8-12)9-11-3-1-2-4-13(11)15/h1-8,14-15H,9H2 |
| SMILES | Oc1ccc(Cc2ccccc2O)cc1 |
| Molecular Weight (g/mol) | 200.23 |
| Formula Weight | 200.0837 |
| Show More |