missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzophenone, TRC

Product Code. 30010209
Change view
Click to view available options
Quantity:
100 mg
1 g
100 g
Unit Size:
100g
100mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010209 100 mg 100mg
30138251 1 g 1g
30132906 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010209 Supplier Toronto Research Chemicals Supplier No. TRCD451780100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-56-6
Formula Weight 214.063
InChI Formula InChI=1S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15H
IUPAC Name (2,4-dihydroxyphenyl)-phenylmethanone
Molecular Formula C13 H10 O3
Molecular Weight (g/mol) 214.22
Percent Purity >95%
SMILES Oc1ccc(C(=O)c2ccccc2)c(O)c1
Synonym Benzophenone, 2,4-dihydroxy- (8CI), (2,4-Dihydroxyphenyl)phenylmethanone, 2,4-Dihydroxybenzophenone, 4-Benzoylresorcinol, ASL 23, Aduvex 12, Advastab 48, Benzophenone 1, Benzoresorcinol, Dastib 263, Eastman inhibitor DHPB, Eversorb 10, HHB, Inhibitor DHBP, Lowilite 24, NC 011, NSC 38555, NSC 5358, Resbenzophenone, Sanduvor 3041, Seesorb 100, Sumisorb 100, Syntase 100, UF 1, UV 0, UV 12, UV 214, UV 214 (UV absorber), UV-O, Ultrafast 800, Uvasorb 20H, Uvinul 3000, Uvinul 400, Uvinul M 400, Uvistat 12, Viosorb 100, Zislizer
Chemical Name or Material 2,4-Dihydroxybenzophenone
Grade HPLC
Show More Show Less

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.