CAS RN 131-56-6
CAS RN 131-56-6
IUPAC Name:
4-benzoylbenzene-1,3-diol
Synonyms:
benzophenone-1 resbenzophenone uvinul 400 HHB dastib 263 2,4-dihydroxybenzofenon uvistat 12 inhibitor dhbp dhbp cpd eastman inhibitor dhpb
Molecular Weight (g/mol):
214.22
Molecular Formula:
C13H10O3
InChi Key:
ZXDDPOHVAMWLBH-UHFFFAOYSA-N
SMILES:
OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
1
–
3
of
3
results
1
2,4-Dihydroxybenzophenone, 99%
CAS: 131-56-6 Molecular Formula: C13H10O3 Molecular Weight (g/mol): 214.22 MDL Number: MFCD00002277 InChI Key: ZXDDPOHVAMWLBH-UHFFFAOYSA-N Synonym: 2,4-dihydroxybenzophenone,2,4-dihydroxyphenyl phenyl methanone,benzophenone-1,benzoresorcinol,resbenzophenone,inhibitor dhbp,uvinul 400,advastab 48,uvistat 12,methanone, 2,4-dihydroxyphenyl phenyl PubChem CID: 8572 ChEBI: CHEBI:34240 IUPAC Name: (2,4-dihydroxyphenyl)-phenylmethanone SMILES: C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O