Learn More
2,4-Dihydroxy-6-amino-5-diethoxyethylpyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxy-6-amino-5-diethoxyethylpyrimidine |
| CAS | 102879-75-4 |
| Molecular Formula | C10 H17 N3 O4 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H17N3O4/c1-3-16-7(17-4-2)5-6-8(11)12-10(15)13-9(6)14/h7H,3-5H2,1-2H3,(H4,11,12,13,14,15) |
| SMILES | CCOC(Cc1c(N)nc(O)nc1O)OCC |
| IUPAC Name | 6-amino-5-(2,2-diethoxyethyl)pyrimidine-2,4-diol |
| Molecular Weight (g/mol) | 243.26 |
| Formula Weight | 243.1219 |