missing translation for 'onlineSavingsMsg'
Learn More

(2,4-difluorophenyl)hydrazine Hydrochloride, TRC

Product Code. 30382227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382227 25 g
30382068 5 g
30372329 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382227 Supplier Toronto Research Chemicals Supplier No. TRCD44824025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40594-30-7
Molecular Formula C6H6F2N2 . HCl
Molecular Weight (g/mol) 144.12
Synonym 1-(2,4-Difluorophenyl)hydrazine Hydrochloride, 2,4-Difluorophenylhydrazine Hydrochloride
SMILES Cl.NNc1ccc(F)cc1F

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H6F2N2/c7-4-1-2-6(10-9)5(8)3-4/h1-3,10H,9H2
Chemical Name or Material (2,4-difluorophenyl)hydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.