Learn More
2,4-Difluorobenzoic Acid, TRC
Product Code. 30136505
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136505 50 g 50g
30134427 25 g 25g
30012538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136505 Supplier Toronto Research Chemicals Supplier No. TRCD44571050G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1583-58-0
Molecular Formula C7 H4 F2 O2
Molecular Weight (g/mol) 158.1
Synonym 2,4-Difluorobenzoic acid, NSC 10312
IUPAC Name 2,4-difluorobenzoic acid
SMILES OC(=O)c1ccc(F)cc1F
Specifications
Quantity 50 g
InChI Formula InChI=1S/C7H4F2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)
Formula Weight 158.0179
Chemical Name or Material 2,4-Difluorobenzoic acid