Learn More
2,4-Diethoxybenzaldehyde, 97%, BLD Pharm™
Product Code. 30948716
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948716 250 mg 250mg
30934246 5 g 5g
30934956 25 g 25g
30930896 100 g 100g
30939528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30948716 Supplier BLD Pharm Supplier No. BD13270250mg
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 22924-16-9
Molecular Formula C11H14O3
MDL Number MFCD00995010
Quantity 250 mg
SMILES O=CC1=CC=C(OCC)C=C1OCC
Molecular Weight (g/mol) 194.23
Percent Purity 97%
Chemical Name or Material 2,4-Diethoxybenzaldehyde