missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrido[3,2-d]pyrimidine, 95%, Thermo Scientific™

Product Code. p-7077456 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15731749 250 mg 250mg
15721749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15731749 Supplier Thermo Scientific Acros Supplier No. 454022500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 39551-54-7
Molecular Formula C7H3Cl2N3
Molecular Weight (g/mol) 200.03
InChI Key UVVFNZJVLRJSMW-UHFFFAOYSA-N
Synonym 2,4-dichloropyrido 3,2-d pyrimidine, pyrido 3,2-d pyrimidine, 2,4-dichloro, pubchem15434, 2,4-dichloro-pyrido 3,2-d pyrimidine, pyrido 3,2-d pyrimidine,2,4-dichloro
PubChem CID 11958053
IUPAC Name 2,4-dichloropyrido[3,2-d]pyrimidine
SMILES C1=CC2=C(C(=NC(=N2)Cl)Cl)N=C1

Specifications

Melting Point 175.0°C to 177.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 200.03
Percent Purity 95%
Chemical Name or Material 2,4-Dichloropyrido[3,2-d]pyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.