missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenoxyacetonitrile, 97%, Thermo Scientific™

Product Code. 15458929
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458929 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458929 Supplier Thermo Scientific Alfa Aesar Supplier No. L10869.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3956-63-6
Melting Point 46°C to 51°C
Molecular Formula C8H5Cl2NO
MDL Number MFCD00019826
Quantity 1 g
UN Number UN3439
Beilstein 1958436
Synonym 2-2,4-dichlorophenoxy acetonitrile, 2,4-dichlorophenoxyacetonitrile, 2,4-dichlorophenoxy acetonitrile, 2-2,4-dichlorophenoxy ethanenitrile, acmc-1coca, maybridge1_008339, 2-2,4-dichloro-phenoxy acetonitrile, 2-2,4-dichlorophenoxy-acetonitrile, 2-2,4-dichloro-phenoxy-acetonitrile, acetonitrile,2-2,4-dichlorophenoxy
InChI Key QSDMPLNZSJLFRM-UHFFFAOYSA-N
SMILES ClC1=CC=C(OCC#N)C(Cl)=C1
IUPAC Name 2-(2,4-dichlorophenoxy)acetonitrile
Molecular Weight (g/mol) 202.03
PubChem CID 245811
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material 2,4-Dichlorophenoxyacetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.