missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenol, Sigma-Aldrich™

Product Code. 15879080
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879080 500 g 500g
15869080 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15879080 Supplier Sigma Aldrich Supplier No. 8037740500
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-83-2
Grade Research
Molecular Formula 2,4-(Cl)2C6H3OH
Molecular Weight (g/mol) 163

Specifications

Melting Point 40°C to 43°C
Boiling Point 209°C to 211°C/1013 hPa
MDL Number MFCD00002169
Quantity 500 g
Synonym 2,4-Dichlorophenol, 2,4-DCP, 2,4-Dichlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.