Learn More
2,4-Dichloro-7-methylquinoline, TRC
Product Code. 30388162
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388162 500 mg
30369250 1 g
30386804 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30388162 Supplier Toronto Research Chemicals Supplier No. TRCD436238500MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102878-19-3
Molecular Formula C10H7NCl2
Molecular Weight (g/mol) 212.08
SMILES Cc1ccc2c(Cl)cc(Cl)nc2c1
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H7Cl2N/c1-6-2-3-7-8(11)5-10(12)13-9(7)4-6/h2-5H,1H3
Formula Weight 210.9956
Chemical Name or Material 2,4-Dichloro-7-methylquinoline