Learn More
2,4-Dichloro-5-thiazolemethanol, TRC
Product Code. 30384829
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384829 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30384829 Supplier Toronto Research Chemicals Supplier No. TRCD48751850MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dichloro-5-thiazolemethanol
CAS 170232-69-6
Molecular Formula C4H3Cl2NOS
Quantity 50 mg
Synonym (2,4-Dichlorothiazol-5-Yl)Methanol
InChI Formula InChI=1S/C4H3Cl2NOS/c5-3-2(1-8)9-4(6)7-3/h8H,1H2
SMILES OCc1sc(Cl)nc1Cl
IUPAC Name (2,4-dichloro-1,3-thiazol-5-yl)methanol
Molecular Weight (g/mol) 184.03
Formula Weight 182.9312