missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-nitroquinazoline, 98%

Product Code. p-7058388
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308745 1 g 1g
11318745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308745 Supplier Thermo Scientific Alfa Aesar Supplier No. L12330.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7154-34-9 | C8H7N5O2 | 205.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7154-34-9
Molecular Formula C8H7N5O2
Molecular Weight (g/mol) 205.18
MDL Number MFCD00023910
InChI Key YZMJNZRTRWPJFY-UHFFFAOYSA-N
Synonym 2,4-diamino-6-nitroquinazoline, 2,4-quinazolinediamine,6-nitro, 6-nitro-2,4-quinazolinediamine #, 2,4-diamino-6-nitro-quinazoline, 1a
PubChem CID 252163
IUPAC Name 6-nitroquinazoline-2,4-diamine
SMILES NC1=NC(N)=C2C=C(C=CC2=N1)[N+]([O-])=O

Specifications

Melting Point >360°C
Quantity 1 g
Beilstein 749531
Formula Weight 205.18
Percent Purity 98%
Chemical Name or Material 2,4-Diamino-6-nitroquinazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.