missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15430946
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430946 250 mg 250mg
15410946 1 g 1g
15420946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430946 Supplier Thermo Scientific Alfa Aesar Supplier No. H54240.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175401-95-3
Molecular Formula C15H9N3O2
Molecular Weight (g/mol) 263.256
MDL Number MFCD20265213
InChI Key HRUUSKALOILBKF-UHFFFAOYSA-N
Synonym 2-4-cyanophenyl benzimidazole-6-carboxylic acid, 2-4-cyanophenyl benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-3h-1,3-benzodiazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-6-carboxylic acid, 1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl, 2-4-cyanophenyl-1h-benzoimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-benzo d imidazole-6-carboxylic acid, 2-4-cyanophenyl-1h-benzimidazole-5-carboxylic acid, 2-4-cyanophenyl-1h-1,3-benzodiazole-5-carboxylic acid
PubChem CID 23509758
IUPAC Name 2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
SMILES C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 263.26
Percent Purity 97%
Chemical Name or Material 2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.