Learn More
2-(4-Chlorophenoxy)ethanol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1892-43-9 |
| Molecular Formula | C8H9ClO2 |
| MDL Number | MFCD00014036 |
| Quantity | 25 g |
| SMILES | ClC1=CC=C(OCCO)C=C1 |
| Molecular Weight (g/mol) | 172.61 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(4-Chlorophenoxy)ethanol |