Learn More
2-[(4-Bromomethyl)phenyl]pyridine, TRC
Product Code. 30339129
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339129 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30339129 Supplier Toronto Research Chemicals Supplier No. TRCB685455100MG
 Restricted Item
Estimated Shipment: 06-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[(4-Bromomethyl)phenyl]pyridine
CAS 52199-24-3
Molecular Formula C12H10BrN
Quantity 100 mg
Synonym 4-(2-Pyridyl)benzyl Bromide
InChI Formula InChI=1S/C12H10BrN/c13-9-10-4-6-11(7-5-10)12-3-1-2-8-14-12/h1-8H,9H2
SMILES BrCc1ccc(cc1)c2ccccn2
IUPAC Name 2-[4-(bromomethyl)phenyl]pyridine
Molecular Weight (g/mol) 248.12
Formula Weight 246.9997