Learn More
2-[(4-Bromomethyl)phenyl]pyridine, TRC
Product Code. 30333142
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333142 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30333142 Supplier Toronto Research Chemicals Supplier No. TRCB6854555G
 Restricted Item
Estimated Shipment: 06-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[(4-Bromomethyl)phenyl]pyridine
CAS 52199-24-3
Molecular Formula C12H10BrN
Quantity 5 g
Synonym 4-(2-Pyridyl)benzyl Bromide
InChI Formula InChI=1S/C12H10BrN/c13-9-10-4-6-11(7-5-10)12-3-1-2-8-14-12/h1-8H,9H2
SMILES BrCc1ccc(cc1)c2ccccn2
IUPAC Name 2-[4-(bromomethyl)phenyl]pyridine
Molecular Weight (g/mol) 248.12
Formula Weight 246.9997