Learn More
2',4',6'-Trimethoxyacetophenone, 98%
Product Code. 11416887
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416887 25 g 25g
11406887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11416887 Supplier Thermo Scientific Alfa Aesar Supplier No. A14328.14
 Restricted Item
In Stock
Add to basket
CAS: 832-58-6 | C11H14O4 | 210.23 g/mol
2',4',6'-Trimethoxyacetophenone is used in the production of 4,2',4',6'-tetramethoxy-chalcone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,4′,6′-Trimethoxyacetophenone is used in the production of 4,2′,4′,6′-tetramethoxy-chalcone.
Solubility
Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 832-58-6
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00017238
InChI Key KPZWHZSIXZXDMW-UHFFFAOYSA-N
Synonym 2',4',6'-trimethoxyacetophenone, 1-2,4,6-trimethoxyphenyl ethanone, 2,4,6-trimethoxyacetophenone, ethanone, 1-2,4,6-trimethoxyphenyl, ethanone,1-2,4,6-trimethoxyphenyl, 1-acetyl-2,4,6-trimethoxybenzene, o-methylxanthoxylin, cambridge id 5135317, 2,4,6-trimethoxy acetophenone, 2/',4/',6/'-trimethoxyacetophenone
PubChem CID 123089
IUPAC Name 1-(2,4,6-trimethoxyphenyl)ethanone
SMILES COC1=CC(OC)=C(C(C)=O)C(OC)=C1
Specifications
Melting Point 101°C to 104°C
Quantity 25 g
Beilstein 519324
Solubility Information Soluble in chloroform.
Formula Weight 210.23
Percent Purity 98%
Chemical Name or Material 2',4',6'-Trimethoxyacetophenone