missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trihydroxyacetophenone Monohydrate, TRC

Product Code. p-200161805
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456527 100 mg -
30449874 50 mg -
30454327 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456527 Supplier Toronto Research Chemicals Supplier No. TRCT888025100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249278-28-2
Molecular Formula C8H8O4 . H2O
Molecular Weight (g/mol) 186.16
Synonym 1-(2,4,6-Trihydroxyphenyl)ethan-1-one Hydrate
IUPAC Name 1-(2,4,6-trihydroxyphenyl)ethanone;hydrate
SMILES O.CC(=O)c1c(O)cc(O)cc1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8O4.H2O/c1-4(9)8-6(11)2-5(10)3-7(8)12;/h2-3,10-12H,1H3;1H2
Formula Weight 186.0528
Chemical Name or Material 2',4',6'-Trihydroxyacetophenone Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.