missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoroaniline, 98%

Product Code. p-7081132
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464393 1 g 1g
11474393 5 g 5g
11484393 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464393 Supplier Thermo Scientific Alfa Aesar Supplier No. A10964.03
Add to basket
This item is not returnable. View return policy

CAS: 363-81-5 | C6H4F3N | 147.1 g/mol

2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N?-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N′-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-81-5
Molecular Formula C6H4F3N
MDL Number MFCD00007650
InChI Key BJSVKBGQDHUBHZ-UHFFFAOYSA-N
Synonym 2-amino-1,3,5-trifluorobenzene, 2,4,6-trifluorobenzenamine, benzenamine, 2,4,6-trifluoro, 2,4,6-trifluorophenylamine, pubchem2468, acmc-1cmhu, 2,4,6 trifluoroaniline, 2,4,6-tri-fluoroaniline, 2,4,6-trifluoro aniline, 2,4,6-trifluoro-aniline
PubChem CID 67765
IUPAC Name 2,4,6-trifluoroaniline

Specifications

Melting Point 33°C to 36°C
Boiling Point 57°C (22 mmHg)
Flash Point 54°C (129°F)
Odor Recognizable
Quantity 1 g
UN Number UN2926
Beilstein 2831022
SMILES C1=C(C=C(C(=C1F)N)F)F
Molecular Weight (g/mol) 147.1
Formula Weight 147.1
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.