missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorophenol, Sigma-Aldrich™

Product Code. 15886570
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886570 100 g 100g
15830431 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15886570 Supplier Sigma-Aldrich Supplier No. 8184690100
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-06-2
Grade Research
Molecular Weight (g/mol) 197.45

Specifications

Melting Point 65°C to 68°C
Density 1.675 g/cm3 at 25°C
Boiling Point 244°C to 246°C/1013 hPa
MDL Number MFCD00002172
Quantity 100 g
Synonym 2,4,6-Trichlorophenol, 2,4,6-Trichlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.