Learn More
2,4,6-Trichlorophenol-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,6-Trichlorophenol-13C6 |
| CAS | 208461-28-3 |
| Molecular Formula | 13C6 H3 Cl3 O |
| Quantity | 25 mg |
| Synonym | Phenol-13C6','2,4,6-trichloro- (9CI), 2,4,6-Trichlorophenol-1,2,3,4,5,6-13C6, 2,4,6-Trichlorophenol 13C6, 2,4,6-Trichlorophenol (ring-13C6) |
| InChI Formula | InChI=1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H/i1+1,2+1,3+1,4+1,5+1,6+1 |
| SMILES | O[13c]1[13c](Cl)[13cH][13c](Cl)[13cH][13c]1Cl |
| IUPAC Name | 2,4,6-trichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol |
| Molecular Weight (g/mol) | 203.4 |
| Formula Weight | 201.9451 |