missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4,6-Tribromopyrimidine, 95%

Product Code. 15403697
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403697 250 mg 250mg
11347349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403697 Supplier Alfa Aesar™ Supplier No. H27471.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 36847-11-7
Molecular Formula C4HBr3N2
Molecular Weight (g/mol) 316.78
MDL Number MFCD00233942
InChI Key AHEYFWKLKMOHCI-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-tribromo, pubchem6896, acmc-1ae09, 2,4,6-tribromopyrimidine
PubChem CID 258580
IUPAC Name 2,4,6-tribromopyrimidine
SMILES BrC1=CC(Br)=NC(Br)=N1

Specifications

Quantity 250 mg
Formula Weight 316.78
Percent Purity 95%
Chemical Name or Material 2,4,6-Tribromopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.