Learn More
2,4,6-Tribromophenol-3,5-d2, TRC
Product Code. 30454800
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454800 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454800 Supplier Toronto Research Chemicals Supplier No. TRCT7726115MG
 Restricted Item
Estimated Shipment: 11-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4,6-Tribromophenol (3,5-D2)
CAS 1219795-42-2
Molecular Formula C6 2H2 H Br3 O
Quantity 5 mg
Synonym Phen-3,5-d2-ol','2,4,6-tribromo-, 2,4,6-Tribromophen-3,5-d2-ol, 2,4,6-Tribromophenol D2, 3,5-Dideuterio-2,4,6-tribromophenol
InChI Formula InChI=1S/C6H3Br3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H/i1D,2D
SMILES [2H]c1c(Br)c([2H])c(Br)c(O)c1Br
IUPAC Name 2,4,6-tribromo-3,5-dideuteriophenol
Molecular Weight (g/mol) 332.81
Formula Weight 329.786