Learn More
2-(4,6-Diphenyl-1,3,5-triazine-2-yl)-5-[(hexyl)oxy]phenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine |
| CAS | 147315-50-2 |
| Molecular Formula | C27 H27 N3 O2 |
| Quantity | 10 g |
| Synonym | 2-(4,6-Diphenyl-1,3,5-triazin-2-yl)-5-(hexyloxy)phenol, 2,4-Diphenyl-6-(2-hydroxy-4-hexyloxyphenyl)-s-triazine, 2,4-Diphenyl-6-[2-hydroxy-4-(hexyloxy)phenyl]-1,3,5-triazine, 2-(2-Hydroxy-4-hexyloxyphenyl)-4,6-diphenyl-1,3,5-triazine, 2-(4,6-Diphenyl-1,3,5-triazine-2-yl)-5-[(hexyl)oxy]phenol, 2-(4,6-Diphenyl-s-triazin-2-yl)-5-hexyloxyphenol, 2-[4-(Hexyloxy)-2-hydroxyphenyl]-4,6-diphenyl-1,3,5-triazine, 4,6-Diphenyl-2-(4-hexyloxy-2-hydroxyphenyl)-s-triazine, Light stabilizer 1577, Songsorb 1577, Tinuvin 1577, Tinuvin 1577ED, Tinuvin 1577FF, Tinuvin 167FF, Tinuvin 577FF, UV 1577 |
| InChI Formula | InChI=1S/C27H27N3O2/c1-2-3-4-11-18-32-22-16-17-23(24(31)19-22)27-29-25(20-12-7-5-8-13-20)28-26(30-27)21-14-9-6-10-15-21/h5-10,12-17,19,31H,2-4,11,18H2,1H3 |
| SMILES | CCCCCCOc1ccc(c(O)c1)c2nc(nc(n2)c3ccccc3)c4ccccc4 |
| IUPAC Name | 2-(4,6-diphenyl-1,3,5-triazin-2-yl)-5-hexoxyphenol |
| Molecular Weight (g/mol) | 425.52 |
| Formula Weight | 425.2103 |