missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethyl-3-oxazoline, cis + trans, 99%, Thermo Scientific™

Product Code. 11495866
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495866 100 g 100g
11475866 5 g 5g
11485866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495866 Supplier Thermo Scientific Alfa Aesar Supplier No. B22228.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4,5-Trimethyl-3-oxazoline, cis + trans possesses the characteristic meat aroma hences used for flavoring application. Used as flavour enhancer in fish

Solubility
Soluble in alcohol, fixed oils. Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22694-96-8
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00053161
InChI Key YFSGRMONVCFYTC-UHFFFAOYSA-N
PubChem CID 31492
IUPAC Name 2,4,5-trimethyl-2,5-dihydro-1,3-oxazole
SMILES CC1C(=NC(O1)C)C

Specifications

Boiling Point 45°C to 47°C (12 mmHg)
Odor Characteristic
Quantity 100 g
UN Number UN1993
Sensitivity Air Sensitive
Synonym 2,4,5-trimethyl-3-oxazoline, 2,4,5-trimethyl-2,5-dihydrooxazole, 2,5-dihydro-2,4,5-trimethyloxazole, 3-oxazoline, 2,4,5-trimethyl, fema no. 3525, oxazole, 2,5-dihydro-2,4,5-trimethyl, 2,4,5-trimethyl-delta-3-oxazoline, oxazole, 2,5-dihydro-2,4,5-trimethl, 2,4,5-trimethyl-3-oxazoline, cis + trans, oxazole,2,5-dihydro-2,4,5-trimethyl
Solubility Information Soluble in alcohol,fixed oils. Insoluble in water.
Formula Weight 113.16
Percent Purity 99%
Chemical Name or Material 2,4,5-Trimethyl-3-oxazoline, cis + trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.