Learn More
2,4,5-Trichlorophenoxyacetic Acid Isobutyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4,5-T-2-methyl-1-propyl ester (mix of isomers) |
| CAS | 4938-72-1 |
| Molecular Formula | C12 H13 Cl3 O3 |
| Quantity | 1 g |
| Synonym | 2,4,5-T-iso-butyl ester |
| InChI Formula | InChI=1S/C12H13Cl3O3/c1-7(2)5-18-12(16)6-17-11-4-9(14)8(13)3-10(11)15/h3-4,7H,5-6H2,1-2H3 |
| SMILES | CC(C)COC(=O)COc1cc(Cl)c(Cl)cc1Cl |
| IUPAC Name | 2-methylpropyl 2-(2,4,5-trichlorophenoxy)acetate |
| Molecular Weight (g/mol) | 311.59 |
| Formula Weight | 309.993 |