Learn More
2-(4,5-Dimethoxy-2-nitrophenyl)acetic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 73357-18-3 |
| Molecular Formula | C10H11NO6 |
| MDL Number | MFCD00118466 |
| Quantity | 5 g |
| SMILES | O=C(O)CC1=CC(OC)=C(OC)C=C1[N+]([O-])=O |
| Molecular Weight (g/mol) | 241.20 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(4,5-Dimethoxy-2-nitrophenyl)acetic acid |