Learn More
2,3-Naphthalic Anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3-Naphthalic Anhydride |
| CAS | 716-39-2 |
| Molecular Formula | C12 H6 O3 |
| Quantity | 5 g |
| Synonym | Naphtho[2,3-c]furan-1,3-dione |
| InChI Formula | InChI=1S/C12H6O3/c13-11-9-5-7-3-1-2-4-8(7)6-10(9)12(14)15-11/h1-6H |
| SMILES | O=C1OC(=O)c2cc3ccccc3cc12 |
| IUPAC Name | benzo[f][2]benzofuran-1,3-dione |
| Molecular Weight (g/mol) | 198.17 |
| Formula Weight | 198.0317 |