Learn More
2-(3-Methoxyphenyl)cyclohexanone, tech. 90%, Thermo Scientific™
Product Code. 11475217
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475217 1 g 1g
11485217 5 g 5g
11495217 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11475217 Supplier Thermo Scientific Alfa Aesar Supplier No. B22619.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15547-89-4
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00001630
InChI Key KACQSVYTBQDRGP-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl cyclohexanone, 2-3-methoxyphenyl cyclohexan-1-one, cyclohexanone, 2-3-methoxyphenyl, acmc-20aoq7, 2-3-methoxyphenyl-cyclohexanone, 2-3-methoxy-phenyl-cyclohexanone, cyclohexanone,2-3-methoxyphenyl, 2-3-methoxyphenyl-cyclohexan-1-one, 2-3-methoxyphenyl cyclohexanone, technical grade
PubChem CID 85868
IUPAC Name 2-(3-methoxyphenyl)cyclohexan-1-one
SMILES COC1=CC=CC(=C1)C2CCCCC2=O
Specifications
Density 1.1
Boiling Point 140°C to 142°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 1 g
Formula Weight 204.27
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone