accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 15547-89-4

IUPAC Name: 2-(3-methoxyphenyl)cyclohexan-1-one
Synonyms: 2-(3-methoxyphenyl)cyclohexan-1-one
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: KACQSVYTBQDRGP-UHFFFAOYNA-N
SMILES: COC1=CC=CC(=C1)C1CCCCC1=O
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)