missing translation for 'onlineSavingsMsg'
Learn More

2’,3’-Dithiouridine, TRC

Product Code. 30380765
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380765 10 mg
30375921 5 mg
30375751 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380765 Supplier Toronto Research Chemicals Supplier No. TRCD49437510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156592-92-6
Molecular Formula C9 H12 N2 O4 S2
Molecular Weight (g/mol) 276.33
IUPAC Name 1-[(2R,3S,4R,5R)-5-(hydroxymethyl)-3,4-bis(sulfanyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](S)[C@@H]1S)N2C=CC(=O)NC2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H12N2O4S2/c12-3-4-6(16)7(17)8(15-4)11-2-1-5(13)10-9(11)14/h1-2,4,6-8,12,16-17H,3H2,(H,10,13,14)/t4-,6-,7-,8-/m1/s1
Formula Weight 276.0238
Chemical Name or Material 2',3'-Dithiouridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.