CAS RN 156592-92-6
CAS RN 156592-92-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2’,3’-Dithiouridine, TRC
CAS: 156592-92-6 Molecular Formula: C9 H12 N2 O4 S2 Molecular Weight (g/mol): 276.33 IUPAC Name: 1-[(2R,3S,4R,5R)-5-(hydroxymethyl)-3,4-bis(sulfanyl)oxolan-2-yl]pyrimidine-2,4-dione SMILES: OC[C@H]1O[C@H]([C@H](S)[C@@H]1S)N2C=CC(=O)NC2=O