Learn More
2,3-Dioxoindoline-7-carboxylic acid, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 25128-35-2 |
| Molecular Formula | C9H5NO4 |
| MDL Number | MFCD00612492 |
| Quantity | 250 mg |
| SMILES | O=C(C1=CC=CC2=C1NC(C2=O)=O)O |
| Molecular Weight (g/mol) | 191.14 |
| Percent Purity | 96% |
| Chemical Name or Material | 2,3-Dioxoindoline-7-carboxylic acid |