Learn More
(2,3-Dimethylpyridin-4-yl)boronic Acid, TRC
Product Code. 30373599
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30373599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30373599 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD4790101G
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material (2,3-Dimethylpyridin-4-yl)boronic Acid
CAS 1246829-05-9
Molecular Formula C7 H10 B N O2
Quantity 1 g
InChI Formula InChI=1S/C7H10BNO2/c1-5-6(2)9-4-3-7(5)8(10)11/h3-4,10-11H,1-2H3
SMILES Cc1nccc(B(O)O)c1C
IUPAC Name (2,3-dimethylpyridin-4-yl)boronic acid
Molecular Weight (g/mol) 150.97
Formula Weight 151.0805