missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpyrazine, TRC

Product Code. 30135293
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135293 5 g 5g
30134316 10 g 10g
30007319 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135293 Supplier Toronto Research Chemicals Supplier No. TRCD4941285G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5910-89-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
Synonym 2,3-Dimethylpyrazine, Pyrazine, 2,3-dimethyl- (6CI, 7CI, 8CI, 9CI, ACI)
SMILES Cc1nccnc1C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H8N2/c1-5-6(2)8-4-3-7-5/h3-4H,1-2H3
Formula Weight 108.0687
Chemical Name or Material 2,3-Dimethylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.