missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpyrazine, 99%

Product Code. 11460463
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460463 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460463 Supplier Thermo Scientific Alfa Aesar Supplier No. A12628.30
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5910-89-4 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5910-89-4
Melting Point -12°C
Density 1.022
Boiling Point 156°C
Flash Point 54°C
Odor Pleasant
Molecular Formula C6H8N2
Refractive Index 1.507
MDL Number MFCD00006144
Quantity 250 g
UN Number UN1993
Beilstein 107908
Synonym pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine
InChI Key OXQOBQJCDNLAPO-UHFFFAOYSA-N
SMILES CC1=NC=CN=C1C
IUPAC Name 2,3-dimethylpyrazine
Molecular Weight (g/mol) 108.14
PubChem CID 22201
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylpyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.