Learn More
2,3-Dimethylpyrazine, 99%
Product Code. 11450463
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450463 50 g 50g
11440463 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11450463 Supplier Thermo Scientific Alfa Aesar Supplier No. A12628.18
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
CAS: 5910-89-4 | C6H8N2 | 108.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5910-89-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006144
InChI Key OXQOBQJCDNLAPO-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dimethyl, 2,3-dimethyl-1,4-diazine, 2,3-dimethyl-pyrazine, pyrazine, dimethyl, 2,3-dimethyl pyrazine, ccris 2928, unii-whf7883d0v, fema no. 3271, natural, dimethylpyrazine
PubChem CID 22201
IUPAC Name 2,3-dimethylpyrazine
SMILES CC1=NC=CN=C1C
Specifications
Melting Point -12°C
Density 1.022
Boiling Point 156°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.507
Quantity 50 g
UN Number UN1993
Beilstein 107908
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,3-Dimethylpyrazine