Learn More
2,3-Dimethylbenzaldehyde, TRC
Specifications
Specifications
| CAS | 5779-93-1 |
| Molecular Formula | C9 H10 O |
| Quantity | 10 g |
| Synonym | Benzaldehyde, 2,3-dimethyl-, 2,3-Dimethylbenzaldehyde, Hemellitaldehyde |
| InChI Formula | InChI=1S/C9H10O/c1-7-4-3-5-9(6-10)8(7)2/h3-6H,1-2H3 |
| SMILES | Cc1cccc(C=O)c1C |
| IUPAC Name | 2,3-dimethylbenzaldehyde |
| Molecular Weight (g/mol) | 134.18 |
| Formula Weight | 134.0732 |
| Chemical Name or Material | 2,3-Dimethylbenzaldehyde |