missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-dimethyl-1h-indole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 10 g |
| Synonym | 1h-indole-5-carboxylic acid, 2,3-dimethyl, 2,3-dimethylindole-5-carboxylic acid, 5-carboxy-2,3-dimethyl-1h-indole, 2,3-dimethyl-indole-5-carboxylic acid, 1h-indole-5-carboxylicacid,2,3-dimethyl, 1h-indole-5-carboxylicacid, 2,3-dimethyl, 2,3-dimethyl-1h-indole-5-carboxylic acid |
| Formula Weight | 189.21 |
| Percent Purity | 97% |
| Chemical Name or Material | 2, 3-Dimethyl-1H-indole-5-carboxylic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction