accessibility menu, dialog, popup

UserName

CAS RN 14844-73-6

H3CHNH3COO

CAS RN 14844-73-6

IUPAC Name: 2,3-dimethyl-1H-indole-5-carboxylate
Synonyms: 2,3-dimethyl-1H-indole-5-carboxylate
Molecular Weight (g/mol): 188.21
Molecular Formula: C11H10NO2
InChi Key: KHJGIMZYCBPOBG-UHFFFAOYSA-M
SMILES: CC1=C(C)C2=CC(=CC=C2N1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results