Learn More
2,3-Dimethoxyaniline, TRC
Product Code. 30385263
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385263 100 mg
30375065 50 mg
30369023 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30385263 Supplier Toronto Research Chemicals Supplier No. TRCD525608100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6299-67-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
Synonym NSC 44873, [2,3-Bis(methyloxy)phenyl]amine, 2,3-Dimethoxybenzenamine
IUPAC Name 2,3-dimethoxyaniline
SMILES O(C1=CC=CC(N)=C1OC)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H11NO2/c1-10-7-5-3-4-6(9)8(7)11-2/h3-5H,9H2,1-2H3
Formula Weight 153.08
Chemical Name or Material 2,3-Dimethoxyaniline