Learn More
2,3-Difluorophenylacetonitrile, 97%, Thermo Scientific™
Product Code. 11428787
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428787 1 g 1g
11438787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428787 Supplier Thermo Scientific Alfa Aesar Supplier No. A19934.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145689-34-5
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00061277
InChI Key IYRCHGRRMKOSHW-UHFFFAOYSA-N
Synonym 2,3-difluorophenylacetonitrile, 2-2,3-difluorophenyl acetonitrile, 2,3-difluorobenzyl cyanide, 2,3-difluorophenylacetanitrile, benzeneacetonitrile, 2,3-difluoro, 2-2,3-difluorophenyl ethanenitrile, 2,3-difluorobenzeneacetonitrile, pubchem1587, acmc-209x0i, ksc183q4t
PubChem CID 518968
IUPAC Name 2-(2,3-difluorophenyl)acetonitrile
SMILES FC1=CC=CC(CC#N)=C1F
Specifications
Refractive Index 1.485
Quantity 1 g
UN Number UN3276
Formula Weight 153.13
Percent Purity 97%
Chemical Name or Material 2,3-Difluorophenylacetonitrile